Anti-RNF20 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RNF20
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-197
- 77829-198
- Antigen Symbol:
- RNF20
- RNF20
- Antigen Name:
- E3 ubiquitin-protein ligase BRE1A protein (1-100AA)
- E3 ubiquitin-protein ligase BRE1A protein (1-100AA)
- Antigen Synonyms:
- BRE1 A| BRE1| BRE1 E3 ubiquitin ligase homolog| BRE1-A| BRE1A| BRE1A_HUMAN| E3 ubiquitin-protein ligase BRE1A| hBRE1| Homolog of S. cerevisiae BRE1| RING finger protein 20| Ring finger protein 20 E3 ubiquitin protein ligase| Rnf20
- BRE1 A| BRE1| BRE1 E3 ubiquitin ligase homolog| BRE1-A| BRE1A| BRE1A_HUMAN| E3 ubiquitin-protein ligase BRE1A| hBRE1| Homolog of S. cerevisiae BRE1| RING finger protein 20| Ring finger protein 20 E3 ubiquitin protein ligase| Rnf20
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5VTR2
- Q5VTR2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together