Anti-API5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: API5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77813-481
- 77813-480
- Antigen Symbol:
- API5
- API5
- Antigen Name:
- Synthetic peptide of API5
- Synthetic peptide of API5
- Antigen Synonyms:
- AAC 11|AAC-11|AAC11|Antiapoptosis clone 11 protein|API 5|API-5|API5 like 1|API5_HUMAN|API5L1|Apoptosis inhibitor 5|Cell migration inducing gene 8 protein|Cell migration-inducing gene 8 protein|Fibroblast growth factor 2 interacting factor 2|Fibroblast growth factor 2-interacting factor|FIF|MIG 8|MIG8|Migration inducing protein MIG8|Protein XAGL
- AAC 11|AAC-11|AAC11|Antiapoptosis clone 11 protein|API 5|API-5|API5 like 1|API5_HUMAN|API5L1|Apoptosis inhibitor 5|Cell migration inducing gene 8 protein|Cell migration-inducing gene 8 protein|Fibroblast growth factor 2 interacting factor 2|Fibroblast growth factor 2-interacting factor|FIF|MIG 8|MIG8|Migration inducing protein MIG8|Protein XAGL
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZZ5
- Q9BZZ5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together