Anti-NAA60 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: NAA60
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77839-498
- 77839-497
- Antigen Symbol:
- NAA60
- NAA60
- Antigen Name:
- N-alpha-acetyltransferase 60 protein (1-94AA)
- N-alpha-acetyltransferase 60 protein (1-94AA)
- Antigen Synonyms:
- FLJ11693| FLJ14154| HAT4| Histone acetyltransferase type B protein 4| N acetyltransferase 15 (GCN5 related, putative)| N alpha acetyltransferase 60| N(alpha) acetyltransferase 60, NatF catalytic subunit| N-acetyltransferase 15| N-alpha-acetyltransferase 60| naa60| NAA60_HUMAN| NAT15| NatF catalytic subunit
- FLJ11693| FLJ14154| HAT4| Histone acetyltransferase type B protein 4| N acetyltransferase 15 (GCN5 related, putative)| N alpha acetyltransferase 60| N(alpha) acetyltransferase 60, NatF catalytic subunit| N-acetyltransferase 15| N-alpha-acetyltransferase 60| naa60| NAA60_HUMAN| NAT15| NatF catalytic subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7X0
- Q9H7X0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together