Anti-SMARCA1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SMARCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-286
- 77800-285
- Antigen Symbol:
- SMARCA1
- SMARCA1
- Antigen Name:
- Probable global transcription activator SNF2L1 protein (500-601AA)
- Probable global transcription activator SNF2L1 protein (500-601AA)
- Antigen Synonyms:
- SMARCA1|SNF2L|SNF2L1|Probable global transcription activator SNF2L1|EC 3.6.4.-|ATP-dependent helicase SMARCA1|Nucleosome-remodeling factor subunit SNF2L|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
- SMARCA1|SNF2L|SNF2L1|Probable global transcription activator SNF2L1|EC 3.6.4.-|ATP-dependent helicase SMARCA1|Nucleosome-remodeling factor subunit SNF2L|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28370
- P28370
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together