Anti-SRRT Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SRRT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-368
- 77840-369
- Antigen Symbol:
- SRRT
- SRRT
- Antigen Name:
- Serrate RNA effector molecule homolog protein (627-876AA)
- Serrate RNA effector molecule homolog protein (627-876AA)
- Antigen Synonyms:
- ARS2 protein|Arsenate resistance protein ARS2|Arsenite resistance protein 2|Arsenite resistance protein|Arsenite-resistance protein 2|ASR2|MGC126427|Serrate|Serrate RNA effector molecule|Serrate RNA effector molecule homolog (Arabidopsis)|Serrate RNA effector molecule homolog|SRRT|SRRT_HUMAN
- ARS2 protein|Arsenate resistance protein ARS2|Arsenite resistance protein 2|Arsenite resistance protein|Arsenite-resistance protein 2|ASR2|MGC126427|Serrate|Serrate RNA effector molecule|Serrate RNA effector molecule homolog (Arabidopsis)|Serrate RNA effector molecule homolog|SRRT|SRRT_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXP5
- Q9BXP5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together