Anti-FGF1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FGF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77809-714
- 77809-715
- Antigen Symbol:
- FGF1
- FGF1
- Antigen Name:
- Fibroblast growth factor 1 protein (16-155AA)
- Fibroblast growth factor 1 protein (16-155AA)
- Antigen Synonyms:
- Acidic fibroblast growth factor|aFGF|Beta endothelial cell growth factor|Beta-endothelial cell growth factor|ECGF|ECGF beta|ECGF-beta|ECGFA|ECGFB|Endothelial Cell Growth Factor alpha|Endothelial Cell Growth Factor beta|FGF 1|FGF alpha|Fgf1|FGF1_HUMAN|FGFA|Fibroblast Growth Factor 1 Acidic|Fibroblast growth factor 1|GLIO703|HBGF 1|HBGF-1|HBGF1|Heparin binding growth factor 1|Heparin binding growth factor 1 precursor|Heparin-binding growth factor 1
- Acidic fibroblast growth factor|aFGF|Beta endothelial cell growth factor|Beta-endothelial cell growth factor|ECGF|ECGF beta|ECGF-beta|ECGFA|ECGFB|Endothelial Cell Growth Factor alpha|Endothelial Cell Growth Factor beta|FGF 1|FGF alpha|Fgf1|FGF1_HUMAN|FGFA|Fibroblast Growth Factor 1 Acidic|Fibroblast growth factor 1|GLIO703|HBGF 1|HBGF-1|HBGF1|Heparin binding growth factor 1|Heparin binding growth factor 1 precursor|Heparin-binding growth factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05230
- P05230
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together