Anti-M6PR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: M6PR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-802
- 77804-801
- Antigen Symbol:
- M6PR
- M6PR
- Antigen Name:
- Cation-dependent mannose-6-phosphate receptor protein (1-277AA)
- Cation-dependent mannose-6-phosphate receptor protein (1-277AA)
- Antigen Synonyms:
- 46 kDa mannose 6 phosphate receptor|46 kDa mannose 6-phosphate receptor|Cation dependent mannose 6 phosphate receptor|Cation dependent mannose 6 phosphate receptor precursor|Cation-dependent mannose-6-phosphate receptor|CD Man 6 P receptor|CD Man-6-P receptor|CD MPR|CD-M6PR|CD-MPR|FLJ32994|M6pr|M6PR protein|Man6PR|Mannose 6 phosphate receptor (cation dependent)|Mannose 6 phosphate receptor|MPR 46|MPR46|MPRD|MPRD_HUMAN|Mr 46,000 Man6PR|Small mannose 6 phosphate receptor|SMPR
- 46 kDa mannose 6 phosphate receptor|46 kDa mannose 6-phosphate receptor|Cation dependent mannose 6 phosphate receptor|Cation dependent mannose 6 phosphate receptor precursor|Cation-dependent mannose-6-phosphate receptor|CD Man 6 P receptor|CD Man-6-P receptor|CD MPR|CD-M6PR|CD-MPR|FLJ32994|M6pr|M6PR protein|Man6PR|Mannose 6 phosphate receptor (cation dependent)|Mannose 6 phosphate receptor|MPR 46|MPR46|MPRD|MPRD_HUMAN|Mr 46,000 Man6PR|Small mannose 6 phosphate receptor|SMPR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20645
- P20645
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together