Anti-RGL2 Rabbit Recombinant Polyclonal Antibody
77797-152
New Product
- Antigen Name:Ral guanine nucleotide dissociation stimulator-like 2 protein (145-328AA)
- Antigen Symbol:RGL2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O15211
- Antigen Synonyms:GDS related protein|HKE1.5|KE1.5|RAB2L|Ral guanine nucleotide dissociation stimulator-like 2|RalGDS-like 2|RalGDS-like factor|Ras-associated protein RAB2L|RGL2|RGL2_HUMAN
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RGL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat