Anti-ANG Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: ANG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77808-861
- 77808-862
- Antigen Symbol:
- ANG
- ANG
- Antigen Name:
- Angiogenin protein (26-147AA)
- Angiogenin protein (26-147AA)
- Antigen Synonyms:
- ALS9|AMYOTROPHIC LATERAL SCLEROSIS|ANG 1|ANG|ANG I|ANG1|ANGI|ANGI_HUMAN|Angiogenin|Angiogenin ribonuclease RNase A family|5|Epididymis luminal protein 168|HEL168|MGC22466|MGC71966|Ribonuclease 5|Ribonuclease RNase A Family 5|RNase 5|RNASE4|RNASE5
- ALS9|AMYOTROPHIC LATERAL SCLEROSIS|ANG 1|ANG|ANG I|ANG1|ANGI|ANGI_HUMAN|Angiogenin|Angiogenin ribonuclease RNase A family|5|Epididymis luminal protein 168|HEL168|MGC22466|MGC71966|Ribonuclease 5|Ribonuclease RNase A Family 5|RNase 5|RNASE4|RNASE5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P03950
- P03950
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together