Anti-MYBPC2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYBPC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-565
- 77823-564
- Antigen Symbol:
- MYBPC2
- MYBPC2
- Antigen Name:
- Myosin-binding protein C, fast-type protein (739-1141AA)
- Myosin-binding protein C, fast-type protein (739-1141AA)
- Antigen Synonyms:
- C protein, skeletal muscle fast isoform|C-protein|Fast MyBP C|Fast MyBP-C|MGC163408|MYBPC|Mybpc2|MYBPCF|Myosin binding protein C, fast type|Myosin-binding protein C, fast-type|MYPC2_HUMAN|skeletal muscle fast isoform
- C protein, skeletal muscle fast isoform|C-protein|Fast MyBP C|Fast MyBP-C|MGC163408|MYBPC|Mybpc2|MYBPCF|Myosin binding protein C, fast type|Myosin-binding protein C, fast-type|MYPC2_HUMAN|skeletal muscle fast isoform
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14324
- Q14324
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together