Anti-POFUT1 Rabbit Polyclonal Antibody
Catalog # 77794-653
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Protein O fucosyltransferase1
- Antigen Symbol:POFUT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H488
- Antigen Synonyms:FUT 12|FUT12|GDP fucose protein O fucosyltransferase 1|GDP-fucose protein O-fucosyltransferase 1|O Fuc T|o fucosyltransferase protein|O FucT 1|O FucT|O FucT1|O FUT|O-FucT-1|OFUT 1|OFUT|OFUT1|OFUT1_HUMAN|Peptide O fucosyltransferase1|Peptide-O-fucosyltransferase 1|Pofut 1|POFUT1|Protein O fucosyltransferase1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: POFUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat