Anti-SNF8 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SNF8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-106
- 77834-105
- Antigen Symbol:
- SNF8
- SNF8
- Antigen Name:
- Vacuolar-sorting protein SNF8 protein (1-258AA)
- Vacuolar-sorting protein SNF8 protein (1-258AA)
- Antigen Synonyms:
- Dot3|ELL associated protein of 30 kDa|ELL-associated protein of 30 kDa|ESCRT-II complex subunit VPS22|hVps22|snf8|SNF8 ESCRTII complex subunit homolog (S cerevisiae)|SNF8_HUMAN|Vacuolar sorting protein SNF8|Vacuolar-sorting protein SNF8|VPS22
- Dot3|ELL associated protein of 30 kDa|ELL-associated protein of 30 kDa|ESCRT-II complex subunit VPS22|hVps22|snf8|SNF8 ESCRTII complex subunit homolog (S cerevisiae)|SNF8_HUMAN|Vacuolar sorting protein SNF8|Vacuolar-sorting protein SNF8|VPS22
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96H20
- Q96H20
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together