Anti-DNAJB6 Rabbit Recombinant Polyclonal Antibody
77806-578
New Product
- Antigen Name:DnaJ homolog subfamily B member 6 protein (208-326AA)
- Antigen Symbol:DNAJB6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75190
- Antigen Synonyms:DJ4|DnaJ|DnaJ (Hsp40) homolog subfamily B member 6|DNAJ B6|DnaJ homolog subfamily B member 6|DnaJ like 2 protein|DNAJB6|DNJB6_HUMAN|Heat shock protein J2|HHDJ1|HSJ-2|HSJ2|LGMD1E|MRJ|MSJ-1|MSJ1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:2000 to 1:10000, IF: 1:50 to 1:200
Type:
Antigen: DNAJB6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse