Anti-CA12 Rabbit Polyclonal Antibody
77809-223
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of CA XII.
- Antigen Symbol:CA12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 μg
- Form:Liquid
- Gene ID:O43570
- Antigen Synonyms:CA 12|CA XII|CA-XII|CAH12_HUMAN|Carbonate dehydratase XII|Carbonic anhydrase 12|Carbonic anhydrase XII|Carbonic dehydratase|CAXII|FLJ20151|HsT18816|T18816|Tumor antigen HOM RCC 3.1.3|Tumor antigen HOM-RCC-3.1.3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: CA12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat