Anti-RDH11 Rabbit Recombinant Polyclonal Antibody
Catalog # 77796-946
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Retinol dehydrogenase 11 protein (22-318AA)
- Antigen Symbol:RDH11
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8TC12
- Antigen Synonyms:Androgen regulated short chain dehydrogenase/reductase 1|Androgen-regulated short-chain dehydrogenase/reductase 1|ARSDR1|AU045252|C85936|CGI 82|FLJ32633|HCBP12|HCV core binding protein|HCV core binding protein HCBP12|HCV core-binding protein HCBP12|MDT1|Prostate short chain dehydrogenase/reductase 1|Prostate short-chain dehydrogenase/reductase 1|PSDR1|RALR1|RDH11_HUMAN|Retinal reductase 1|retinol dehydrogenase 11 (all trans/9 cis/11 cis)|Retinol dehydrogenase 11|SCALD|SDR7C1|Short chain dehydrogenase/reductase family 7C|member 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: RDH11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human