Anti-CDC25B Rabbit Polyclonal Antibody
Catalog # 77812-486
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from Cdc25B around the phosphorylation site of S353.
- Antigen Symbol:CDC25B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Monkey
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P30305
- Antigen Synonyms:AI604853|Cdc 25B|Cdc25b|CDC25HU2|Cdc25m2|Cell division cycle 25 homolog B|Cell division cycle 25B|Cell division cycle 25B isoform 1|Cell division cycle 25B isoform 2|Cell division cycle 25B isoform 3|Cell division cycle 25B isoform 4|Cell division cycle 25B isoform 5|Dual specificity phosphatase Cdc25B|EC 3.1.3.48|M phase inducer phosphatase 2|M-phase inducer phosphatase 2|MPIP2_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: CDC25B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey