Anti-dsdA Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: dsdA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Salmonella heidelberg
Specifications
- Catalog No:
- 77818-854
- 77818-855
- Antigen Symbol:
- dsdA
- dsdA
- Antigen Name:
- Recombinant Salmonella heidelberg D-serine dehydratase protein (1-440AA)
- Recombinant Salmonella heidelberg D-serine dehydratase protein (1-440AA)
- Antigen Synonyms:
- dsdA| SeHA_C4132| D-serine dehydratase| EC 4.3.1.18| D-serine deaminase| DSD
- dsdA| SeHA_C4132| D-serine dehydratase| EC 4.3.1.18| D-serine deaminase| DSD
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Salmonella heidelberg
- Salmonella heidelberg
- Host:
- Rabbit
- Rabbit
- Gene ID:
- B4TA53
- B4TA53
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together