Anti-EPAS1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: EPAS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-128
- 77835-129
- Antigen Symbol:
- EPAS1
- EPAS1
- Antigen Name:
- Endothelial PAS domain-containing protein 1 protein (543-870AA)
- Endothelial PAS domain-containing protein 1 protein (543-870AA)
- Antigen Synonyms:
- Basic helix loop helix PAS protein MOP2|Basic-helix-loop-helix-PAS protein MOP2|bHLHe73|Class E basic helix-loop-helix protein 73|ECYT4|Endothelial PAS domain containing protein 1|Endothelial pas domain protein 1|Endothelial PAS domain-containing protein 1|EPAS 1|EPAS-1|EPAS1|EPAS1_HUMAN|HIF 1 alpha like factor|HIF 2 alpha|HIF-1-alpha-like factor|HIF-2-alpha|HIF2-alpha|HIF2A|HLF|Hypoxia inducible factor 2 alpha|Hypoxia inducible factor 2 alpha subunit|Hypoxia-inducible factor 2-alpha|Member of PAS protein 2|Member of pas superfamily 2|MOP 2|MOP2|PAS domain-containing protein 2|PASD2
- Basic helix loop helix PAS protein MOP2|Basic-helix-loop-helix-PAS protein MOP2|bHLHe73|Class E basic helix-loop-helix protein 73|ECYT4|Endothelial PAS domain containing protein 1|Endothelial pas domain protein 1|Endothelial PAS domain-containing protein 1|EPAS 1|EPAS-1|EPAS1|EPAS1_HUMAN|HIF 1 alpha like factor|HIF 2 alpha|HIF-1-alpha-like factor|HIF-2-alpha|HIF2-alpha|HIF2A|HLF|Hypoxia inducible factor 2 alpha|Hypoxia inducible factor 2 alpha subunit|Hypoxia-inducible factor 2-alpha|Member of PAS protein 2|Member of pas superfamily 2|MOP 2|MOP2|PAS domain-containing protein 2|PASD2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99814
- Q99814
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together