Anti-SPAG5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SPAG5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77833-209
- 77833-210
- Antigen Symbol:
- SPAG5
- SPAG5
- Antigen Name:
- Sperm-associated antigen 5 protein (1-300AA)
- Sperm-associated antigen 5 protein (1-300AA)
- Antigen Synonyms:
- Astrin|Deepest|hMAP 126|hMAP126|MAP 126|MAP126|Mitotic spindle associated protein|Mitotic spindle associated protein p126|Mitotic spindle coiled coil related protein|Mitotic spindle-associated protein p126|SPAG 5|SPAG5|SPAG5_HUMAN|Sperm associated antigen 5|Sperm tail protein Spag 5|Sperm tail protein Spag5|Sperm-associated antigen 5
- Astrin|Deepest|hMAP 126|hMAP126|MAP 126|MAP126|Mitotic spindle associated protein|Mitotic spindle associated protein p126|Mitotic spindle coiled coil related protein|Mitotic spindle-associated protein p126|SPAG 5|SPAG5|SPAG5_HUMAN|Sperm associated antigen 5|Sperm tail protein Spag 5|Sperm tail protein Spag5|Sperm-associated antigen 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96R06
- Q96R06
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together