Anti-COQ7 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: COQ7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
- Catalog No:
- 77834-750
- 77834-749
- Antigen Symbol:
- COQ7
- COQ7
- Antigen Name:
- 5-demethoxyubiquinone hydroxylase, mitochondrial protein (37-217AA)
- 5-demethoxyubiquinone hydroxylase, mitochondrial protein (37-217AA)
- Antigen Synonyms:
- COQ7|5-demethoxyubiquinone hydroxylase|mitochondrial|DMQ hydroxylase|EC 1.14.13.-|Timing protein clk-1 homolog|Ubiquinone biosynthesis monooxygenase COQ7
- COQ7|5-demethoxyubiquinone hydroxylase|mitochondrial|DMQ hydroxylase|EC 1.14.13.-|Timing protein clk-1 homolog|Ubiquinone biosynthesis monooxygenase COQ7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99807
- Q99807
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together