Anti-TMEM27 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMEM27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-785
- 77840-786
- Antigen Symbol:
- TMEM27
- TMEM27
- Antigen Name:
- Collectrin protein (15-141AA)
- Collectrin protein (15-141AA)
- Antigen Synonyms:
- UNQ679|Collectrin|kidney-specific membrane protein|NX-17|NX17|PRO1312|TMM27_HUMAN|Transmembrane protein 27|UNQ679/PRO1312
- UNQ679|Collectrin|kidney-specific membrane protein|NX-17|NX17|PRO1312|TMM27_HUMAN|Transmembrane protein 27|UNQ679/PRO1312
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBJ8
- Q9HBJ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together