Anti-SFTPA1 Rabbit Polyclonal Antibody
Catalog # 77799-375
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:surfactant-associated protein, pulmonary 1
- Antigen Symbol:SFTPA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IWL2
- Antigen Synonyms:35 kDa pulmonary surfactant associated protein|35 kDa pulmonary surfactant-associated protein|Alveolar proteinosis protein|COLEC4|Collectin 4|Collectin-4|FLJ50593|FLJ51913|FLJ61144|FLJ77898,|FLJ79095|FLJ99559|MGC133365|MGC198590|OTTHUMP00000019928|OTTHUMP00000019929|OTTHUMP00000019930|OTTHUMP00000019931|PSAP|PSP A|PSP-A|PSPA|pulmonary surfactant -associated protein, 35-KD|pulmonary surfactant apoprotein PSP-A|pulmonary surfactant associated protein A1|Pulmonary surfactant-associated protein A1|SFTA1_HUMAN|SFTP1|SFTPA1|SFTPA1B|SP A|SP A1|SP-A|SP-A1|SPA|SPA1|surfactant protein A1|surfactant protein A1 variant AB'D' 6A2|surfactant protein A1 variant AB'D' 6A3|surfactant protein A1 variant AB'D' 6A4|surfactant protein A1 variant ACD' 6A2|surfactant protein A1 variant ACD' 6A3|surfactant protein A1 variant ACD' 6A4|surfactant protein A1 variant AD' 6A|surfactant protein A1 variant AD' 6A2|surfactant protein A1 variant AD' 6A3|surfactant protein A1 variant AD' 6A4|surfactant protein A1B|surfactant, pulmonary associated protein A1A|surfactant, pulmonary associated protein A1B|surfactant-associated protein, pulmonary 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human