Anti-FKBP1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-055
- 77802-054
- Antigen Symbol:
- FKBP1A
- FKBP1A
- Antigen Name:
- Peptidyl-prolyl cis-trans isomerase FKBP1A protein (2-103AA)
- Peptidyl-prolyl cis-trans isomerase FKBP1A protein (2-103AA)
- Antigen Synonyms:
- 12 kDa FK506-binding protein|12 kDa FKBP|Calstabin 1|FK506 binding protein 1|FK506 binding protein 12|FK506 binding protein 1A (12kD)|FK506 binding protein 1A 12kDa|FK506 binding protein 1A|FK506 binding protein T cell 12 kD|FK506 binding protein|T cell|12 kD|FK506 binding protein12|FK506-binding protein 1A|FKB1A_HUMAN|FKBP 12|FKBP 1A|FKBP-12|FKBP-1A|FKBP1|FKBP12|FKBP12 Exip3|FKBP12C|fkbp1a|Immunophilin FKBP12|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP1A|PKC12|PKCI2|PPIase|PPIase FKBP1A|Protein kinase C inhibitor 2|Rotamase
- 12 kDa FK506-binding protein|12 kDa FKBP|Calstabin 1|FK506 binding protein 1|FK506 binding protein 12|FK506 binding protein 1A (12kD)|FK506 binding protein 1A 12kDa|FK506 binding protein 1A|FK506 binding protein T cell 12 kD|FK506 binding protein|T cell|12 kD|FK506 binding protein12|FK506-binding protein 1A|FKB1A_HUMAN|FKBP 12|FKBP 1A|FKBP-12|FKBP-1A|FKBP1|FKBP12|FKBP12 Exip3|FKBP12C|fkbp1a|Immunophilin FKBP12|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP1A|PKC12|PKCI2|PPIase|PPIase FKBP1A|Protein kinase C inhibitor 2|Rotamase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62942
- P62942
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together