Anti-MAPRE1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: MAPRE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77824-126
- 77824-125
- Antigen Symbol:
- 5530600P05Rik;Adenomatous polyposis coli binding protein EB 1;Adenomatous polyposis coli binding protein EB1;AI462499;AI504412;APC binding protein EB 1;APC binding protein EB1;APC-binding protein EB1;AW260097;BIM1p;D2Ertd459e;EB 1;EB1;End binding protein 1;End-binding protein 1;fa01e12;fc23e11;fi33c06;MAPRE 1;MAPRE1;Mapre3;MARE1_HUMAN;MGC117374;MGC129946;MGC52508;Microtubule associated protein RP/EB family member 1;Microtubule-associated protein RP/EB family member 1;wu:fa01e12;wu:fc23e11;wu:fi33c06;zgc:55428;zgc:77807;zgc:85755
- 5530600P05Rik;Adenomatous polyposis coli binding protein EB 1;Adenomatous polyposis coli binding protein EB1;AI462499;AI504412;APC binding protein EB 1;APC binding protein EB1;APC-binding protein EB1;AW260097;BIM1p;D2Ertd459e;EB 1;EB1;End binding protein 1;End-binding protein 1;fa01e12;fc23e11;fi33c06;MAPRE 1;MAPRE1;Mapre3;MARE1_HUMAN;MGC117374;MGC129946;MGC52508;Microtubule associated protein RP/EB family member 1;Microtubule-associated protein RP/EB family member 1;wu:fa01e12;wu:fc23e11;wu:fi33c06;zgc:55428;zgc:77807;zgc:85755
- Antigen Name:
- Microtubule-associated protein RP/EB family member 1 protein (2-268AA)
- Microtubule-associated protein RP/EB family member 1 protein (2-268AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15691
- Q15691
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together