Order Entry
United States
ContactUsLinkComponent
Anti-CCDC68 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-CCDC68 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CCDC68 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77840-581
  • 77840-582
  • Antigen Symbol:
  • CCDC68
  • CCDC68
  • Antigen Name:
  • Coiled-coil domain-containing protein 68 protein (131-335AA)
  • Coiled-coil domain-containing protein 68 protein (131-335AA)
  • Antigen Synonyms:
  • CCD68_HUMAN|CCDC 68|CCDC68|Coiled coil domain containing 68|Coiled coil domain containing protein 68|Coiled-coil domain-containing protein 68|CTCL associated antigen se57 1|CTCL tumor antigen se57 1|CTCL-associated antigen se57-1|Cutaneous T cell lymphoma associated antigen|Cutaneous T cell lymphoma associated antigen se57 1|Cutaneous T-cell lymphoma-associated antigen se57-1|FLJ25368|SE57 1
  • CCD68_HUMAN|CCDC 68|CCDC68|Coiled coil domain containing 68|Coiled coil domain containing protein 68|Coiled-coil domain-containing protein 68|CTCL associated antigen se57 1|CTCL tumor antigen se57 1|CTCL-associated antigen se57-1|Cutaneous T cell lymphoma associated antigen|Cutaneous T cell lymphoma associated antigen se57 1|Cutaneous T-cell lymphoma-associated antigen se57-1|FLJ25368|SE57 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H2F9
  • Q9H2F9
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together