Anti-CELSR2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: CELSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77808-499
- 77808-500
- Antigen Symbol:
- CELSR2
- CELSR2
- Antigen Name:
- Synthetic peptide of CELSR2
- Synthetic peptide of CELSR2
- Antigen Synonyms:
- Cadherin EGF LAG seven pass G type receptor 2 flamingo Drosophila homolog|Cadherin EGF LAG seven-pass G-type receptor 2|Cadherin family member 10|CDHF10|CELR2_HUMAN|EGF like domain multiple 2|EGF-like protein 2|EGFL2|epidermal growth factor like 2|Epidermal growth factor-like protein 2|Flamingo homolog 3|Flamingo1|KIAA0279|MEGF3|Multiple EGF-like domains protein 3|Multiple epidermal growth factor-like domains 3|Multiple epidermal growth factor-like domains protein 3
- Cadherin EGF LAG seven pass G type receptor 2 flamingo Drosophila homolog|Cadherin EGF LAG seven-pass G-type receptor 2|Cadherin family member 10|CDHF10|CELR2_HUMAN|EGF like domain multiple 2|EGF-like protein 2|EGFL2|epidermal growth factor like 2|Epidermal growth factor-like protein 2|Flamingo homolog 3|Flamingo1|KIAA0279|MEGF3|Multiple EGF-like domains protein 3|Multiple epidermal growth factor-like domains 3|Multiple epidermal growth factor-like domains protein 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCU4
- Q9HCU4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together