Anti-WIZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WIZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-607
- 77804-606
- Antigen Symbol:
- WIZ
- WIZ
- Antigen Name:
- Protein Wiz protein (1469-1574AA)
- Protein Wiz protein (1469-1574AA)
- Antigen Synonyms:
- Protein Wiz|Widely interspaced zinc finger containing protein|Widely-interspaced zinc finger-containing protein|WIZ zinc finger|WIZ_HUMAN|Zinc finger protein 803|ZNF803
- Protein Wiz|Widely interspaced zinc finger containing protein|Widely-interspaced zinc finger-containing protein|WIZ zinc finger|WIZ_HUMAN|Zinc finger protein 803|ZNF803
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95785
- O95785
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together