Anti-POLR3F Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLR3F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-960
- 77841-961
- Antigen Symbol:
- POLR3F
- POLR3F
- Antigen Name:
- DNA-directed RNA polymerase III subunit RPC6 protein (1-270AA)
- DNA-directed RNA polymerase III subunit RPC6 protein (1-270AA)
- Antigen Synonyms:
- DNA directed RNA polymerase III 39 kDa polypeptide|DNA directed RNA polymerase III 39 kDa polypeptide F|DNA directed RNA polymerase III subunit F|DNA directed RNA polymerase III subunit RPC6|DNA directed RNA polymerases III 39 kDa polypeptide|DNA-directed RNA polymerase III subunit F|DNA-directed RNA polymerase III subunit RPC6|MGC13517|Polr3f|Polymerase (RNA) III (DNA directed) polypeptide F|RNA polymerase III 39 kDa subunit|RNA polymerase III C39 subunit|RNA polymerase III subunit C6|RPC 39|RPC 6|RPC39|RPC6|RPC6_HUMAN
- DNA directed RNA polymerase III 39 kDa polypeptide|DNA directed RNA polymerase III 39 kDa polypeptide F|DNA directed RNA polymerase III subunit F|DNA directed RNA polymerase III subunit RPC6|DNA directed RNA polymerases III 39 kDa polypeptide|DNA-directed RNA polymerase III subunit F|DNA-directed RNA polymerase III subunit RPC6|MGC13517|Polr3f|Polymerase (RNA) III (DNA directed) polypeptide F|RNA polymerase III 39 kDa subunit|RNA polymerase III C39 subunit|RNA polymerase III subunit C6|RPC 39|RPC 6|RPC39|RPC6|RPC6_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H1D9
- Q9H1D9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together