Anti-SLC25A11 Rabbit Polyclonal Antibody
Catalog # 77813-236
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of SLC25A11
- Antigen Symbol:SLC25A11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q02978
- Antigen Synonyms:M2OM_HUMAN|Mitochondrial 2 oxoglutarate/malate carrier protein|Mitochondrial 2-oxoglutarate/malate carrier protein|OGC|OGCP|SLC20A4|Solute carrier family 20 (oxoglutarate carrier) member 4|Solute carrier family 20 member 4|Solute carrier family 25 (mitochondrial carrier oxoglutarate carrier) member 11|Solute carrier family 25 member 11
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: SLC25A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human