Anti-RCN2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RCN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-395
- 77824-394
- Antigen Symbol:
- RCN2
- RCN2
- Antigen Name:
- Reticulocalbin-2 protein (23-180AA)
- Reticulocalbin-2 protein (23-180AA)
- Antigen Synonyms:
- Calcium binding protein ERC 55|Calcium-binding protein ERC-55|E6 binding protein|E6-binding protein|E6BP|ERC 55|ERC55|Rcn2|RCN2_HUMAN|Reticulocalbin 2 EF hand calcium binding domain (endoplasmic reticulum calcium binding protein 55kD)|Reticulocalbin 2|EF hand calcium binding domain|Reticulocalbin-2|TCBP49
- Calcium binding protein ERC 55|Calcium-binding protein ERC-55|E6 binding protein|E6-binding protein|E6BP|ERC 55|ERC55|Rcn2|RCN2_HUMAN|Reticulocalbin 2 EF hand calcium binding domain (endoplasmic reticulum calcium binding protein 55kD)|Reticulocalbin 2|EF hand calcium binding domain|Reticulocalbin-2|TCBP49
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14257
- Q14257
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together