Anti-RFPL2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77797-087
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ret finger protein-like 2 protein (99-378AA)
- Antigen Symbol:RFPL2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O75678
- Antigen Synonyms:Ret finger protein like 2|Ret finger protein-like 2|RFPL 2|RFPL2|RFPL2_HUMAN|RING finger protein 79|RNF79
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: RFPL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human