Anti-CCT7 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CCT7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-755
- 77834-754
- Antigen Symbol:
- CCT7
- CCT7
- Antigen Name:
- T-complex protein 1 subunit eta protein (240-543AA)
- T-complex protein 1 subunit eta protein (240-543AA)
- Antigen Synonyms:
- CCT ETA|CCT-eta|CCTETA|Ccth|Chaperonin containing t complex polypeptide 1 eta subunit|Chaperonin containing TCP1 subunit 7 (eta)|Chaperonin containing TCP1 subunit 7|HIV 1 Nef interacting protein|HIV-1 Nef-interacting protein|Nip7 1|T complex protein 1 eta subunit|T-complex protein 1 subunit eta, N-terminally processed|TCP-1-eta|TCP1eta|TCPH_HUMAN
- CCT ETA|CCT-eta|CCTETA|Ccth|Chaperonin containing t complex polypeptide 1 eta subunit|Chaperonin containing TCP1 subunit 7 (eta)|Chaperonin containing TCP1 subunit 7|HIV 1 Nef interacting protein|HIV-1 Nef-interacting protein|Nip7 1|T complex protein 1 eta subunit|T-complex protein 1 subunit eta, N-terminally processed|TCP-1-eta|TCP1eta|TCPH_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99832
- Q99832
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together