Anti-EIF2S1 Rabbit Polyclonal Antibody
Catalog # 77813-893
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:eIF2a
- Antigen Symbol:EIF2S1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P05198
- Antigen Synonyms:EIF 2 alpha|EIF 2|EIF 2A|EIF 2alpha|eIF-2-alpha|eIF-2A|EIF-2alpha|EIF2 alpha|EIF2|EIF2A|EIF2S1|Eukaryotic translation initiation factor 2 subunit 1 alpha 35kDa|Eukaryotic translation initiation factor 2 subunit 1 alpha|Eukaryotic translation initiation factor 2 subunit 1|Eukaryotic translation initiation factor 2 subunit alpha|IF2A_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:200
Type:
Antigen: EIF2S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat