Anti-DUSP26 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DUSP26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-867
- 77835-868
- Antigen Symbol:
- DUSP26
- DUSP26
- Antigen Name:
- Dual specificity protein phosphatase 26 protein (1-211AA)
- Dual specificity protein phosphatase 26 protein (1-211AA)
- Antigen Synonyms:
- DSP-4| Dual specificity phosphatase 26 (putative)| Dual specificity phosphatase 26| Dual specificity phosphatase SKRP3| Dual specificity protein phosphatase 26| DUS26_HUMAN| DUSP24| DUSP26| Hypothetical protein FLJ31142| LDP 4| LDP-4| Low molecular mass dual specificity phosphatase 4| Low-molecular-mass dual-specificity phosphatase 4| MAP kinase phosphatase 8| MGC1136| MGC2627| Mitogen activated protein kinase phosphatase 8| Mitogen-activated protein kinase phosphatase 8| MKP-8| MKP8| NATA1| NATA1 protein| Novel amplified gene in thyroid anaplastic cancer| SKRP3
- DSP-4| Dual specificity phosphatase 26 (putative)| Dual specificity phosphatase 26| Dual specificity phosphatase SKRP3| Dual specificity protein phosphatase 26| DUS26_HUMAN| DUSP24| DUSP26| Hypothetical protein FLJ31142| LDP 4| LDP-4| Low molecular mass dual specificity phosphatase 4| Low-molecular-mass dual-specificity phosphatase 4| MAP kinase phosphatase 8| MGC1136| MGC2627| Mitogen activated protein kinase phosphatase 8| Mitogen-activated protein kinase phosphatase 8| MKP-8| MKP8| NATA1| NATA1 protein| Novel amplified gene in thyroid anaplastic cancer| SKRP3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV47
- Q9BV47
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together