Anti-DHX32 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DHX32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-218
- 77830-217
- Antigen Symbol:
- DHX32
- DHX32
- Antigen Name:
- Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA)
- Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA)
- Antigen Synonyms:
- 3110079L04Rik|AA408140|Ddx32|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32|DEAD/H box 32|DEAD/H helicase like protein 1|DEAD/H helicase-like protein 1|DEAH (Asp-Glu-Ala-His) box polypeptide 32|DEAH box protein 32|DHLP1|DHX32|DHX32_HUMAN|Helicase DDX32|HuDDX32|Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
- 3110079L04Rik|AA408140|Ddx32|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32|DEAD/H box 32|DEAD/H helicase like protein 1|DEAD/H helicase-like protein 1|DEAH (Asp-Glu-Ala-His) box polypeptide 32|DEAH box protein 32|DHLP1|DHX32|DHX32_HUMAN|Helicase DDX32|HuDDX32|Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L7V1
- Q7L7V1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together