Anti-DLEU7 Rabbit Polyclonal Antibody
77832-894
New Product
- Antigen Name:Synthetic peptide from protein at AA range: 51-100
- Antigen Symbol:DLEU7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q6UYE1
- Antigen Synonyms:Deleted in lymphocytic leukemia 7|LEU7_HUMAN|Leukemia-associated protein 7
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:50 to 300, ELISA: 1:10000 to 20000
Type:
Antigen: DLEU7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human