Anti-NEK5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NEK5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-380
- 77829-381
- Antigen Symbol:
- NEK5
- NEK5
- Antigen Name:
- Serine/threonine-protein kinase Nek5 protein (372-468AA)
- Serine/threonine-protein kinase Nek5 protein (372-468AA)
- Antigen Synonyms:
- NEK5|Serine/threonine-protein kinase Nek5|EC 2.7.11.1|Never in mitosis A-related kinase 5|NimA-related protein kinase 5
- NEK5|Serine/threonine-protein kinase Nek5|EC 2.7.11.1|Never in mitosis A-related kinase 5|NimA-related protein kinase 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P3R8
- Q6P3R8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together