Anti-MTMR12 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MTMR12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-460
- 77840-461
- Antigen Symbol:
- MTMR12
- MTMR12
- Antigen Name:
- Myotubularin-related protein 12 protein (188-318AA)
- Myotubularin-related protein 12 protein (188-318AA)
- Antigen Synonyms:
- MTMR12|KIAA1682|PIP3AP|Myotubularin-related protein 12|Inactive phosphatidylinositol 3-phosphatase 12|Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit|3-PAP|3-phosphatase adapter protein
- MTMR12|KIAA1682|PIP3AP|Myotubularin-related protein 12|Inactive phosphatidylinositol 3-phosphatase 12|Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit|3-PAP|3-phosphatase adapter protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C0I1
- Q9C0I1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together