Anti-SH3BGR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3BGR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-451
- 77799-452
- Antigen Symbol:
- SH3BGR
- SH3BGR
- Antigen Name:
- SH3 domain-binding glutamic acid-rich protein (1-239AA)
- SH3 domain-binding glutamic acid-rich protein (1-239AA)
- Antigen Synonyms:
- SH3BGR|SH3 domain-binding glutamic acid-rich protein|SH3BGR protein|21-glutamic acid-rich protein|21-GARP
- SH3BGR|SH3 domain-binding glutamic acid-rich protein|SH3BGR protein|21-glutamic acid-rich protein|21-GARP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55822
- P55822
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together