Anti-KCNIP4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: KCNIP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-333
- 77830-332
- Antigen Symbol:
- KCNIP4
- KCNIP4
- Antigen Name:
- Kv channel-interacting protein 4 protein (1-250AA)
- Kv channel-interacting protein 4 protein (1-250AA)
- Antigen Synonyms:
- A type potassium channel modulatory protein 4| A-type potassium channel modulatory protein 4| CALP| Calsenilin like protein| Calsenilin-like protein| HGNC:30083| KCHIP 4| KChIP4| KCIP4_HUMAN| KCNIP 4| Kcnip4| Kv channel interacting protein 4| Kv channel-interacting protein 4| MGC44947| Potassium channel interacting protein 4| Potassium channel-interacting protein 4
- A type potassium channel modulatory protein 4| A-type potassium channel modulatory protein 4| CALP| Calsenilin like protein| Calsenilin-like protein| HGNC:30083| KCHIP 4| KChIP4| KCIP4_HUMAN| KCNIP 4| Kcnip4| Kv channel interacting protein 4| Kv channel-interacting protein 4| MGC44947| Potassium channel interacting protein 4| Potassium channel-interacting protein 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6PIL6
- Q6PIL6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together