Anti-SRMS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: SRMS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-544
- 77835-543
- Antigen Symbol:
- SRMS
- SRMS
- Antigen Name:
- Tyrosine-protein kinase Srms protein (19-155AA)
- Tyrosine-protein kinase Srms protein (19-155AA)
- Antigen Synonyms:
- C20orf148|dJ697K14.1|EC 2.7.1.112|Src related kinase lacking C terminal regulatory tyrosine and N terminal myristylation sites|SRM|SRMS|SRMS_HUMAN|Tyrosine protein kinase Srms|Tyrosine-protein kinase Srms
- C20orf148|dJ697K14.1|EC 2.7.1.112|Src related kinase lacking C terminal regulatory tyrosine and N terminal myristylation sites|SRM|SRMS|SRMS_HUMAN|Tyrosine protein kinase Srms|Tyrosine-protein kinase Srms
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3Y6
- Q9H3Y6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together