Anti-RAB5A Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: RAB5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77796-451
- 77796-452
- Antigen Symbol:
- RAB5A
- RAB5A
- Antigen Name:
- Ras-related protein Rab-5A protein (1-215AA)
- Ras-related protein Rab-5A protein (1-215AA)
- Antigen Synonyms:
- RAB 5|RAB 5A|RAB5A|RAB5A member RAS oncogene family|RAB5A_HUMAN|RAS associated protein RAB5A|Ras related protein Rab 5A|Ras-related protein Rab-5A
- RAB 5|RAB 5A|RAB5A|RAB5A member RAS oncogene family|RAB5A_HUMAN|RAS associated protein RAB5A|Ras related protein Rab 5A|Ras-related protein Rab-5A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20339
- P20339
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together