Anti-AGPAT3 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: AGPAT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-478
- 77836-479
- Antigen Symbol:
- AGPAT3
- AGPAT3
- Antigen Name:
- 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma protein (148-304AA)
- 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma protein (148-304AA)
- Antigen Synonyms:
- 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma|1-acylglycerol-3-phosphate O-acyltransferase 3|1-AGP acyltransferase 3|1-AGPAT 3|acyltransferase gamma|AGPAT3|LPAAT-gamma|Lysophosphatidic acid acyltransferase gamma|PLCC_HUMAN|UNQ759/PRO1490
- 1-acyl-sn-glycerol-3-phosphate acyltransferase gamma|1-acylglycerol-3-phosphate O-acyltransferase 3|1-AGP acyltransferase 3|1-AGPAT 3|acyltransferase gamma|AGPAT3|LPAAT-gamma|Lysophosphatidic acid acyltransferase gamma|PLCC_HUMAN|UNQ759/PRO1490
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRZ7
- Q9NRZ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together