Anti-ACAA2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: ACAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77822-538
- 77822-537
- Antigen Symbol:
- ACAA2
- ACAA2
- Antigen Name:
- Fusion protein of ACAA2
- Fusion protein of ACAA2
- Antigen Synonyms:
- 3 ketoacyl CoA thiolase|3 ketoacyl CoA thiolase mitochondrial|3-ketoacyl-CoA thiolase, mitochondrial|3-oxoacyl-CoA thiolase, mitochondrial|ACAA 2|Acaa2|Acetyl CoA acyltransferase 2|Acetyl CoA acyltransferase|Acetyl Coenzyme A acyltransferase 2|Acetyl-CoA acyltransferase|Beta ketothiolase|Beta-ketothiolase|DSAEC|Mitochondrial 3 oxoacyl CoA thiolase|Mitochondrial 3 oxoacyl Coenzyme A thiolase|Mitochondrial 3-oxoacyl-CoA thiolase|T1|THIM_HUMAN
- 3 ketoacyl CoA thiolase|3 ketoacyl CoA thiolase mitochondrial|3-ketoacyl-CoA thiolase, mitochondrial|3-oxoacyl-CoA thiolase, mitochondrial|ACAA 2|Acaa2|Acetyl CoA acyltransferase 2|Acetyl CoA acyltransferase|Acetyl Coenzyme A acyltransferase 2|Acetyl-CoA acyltransferase|Beta ketothiolase|Beta-ketothiolase|DSAEC|Mitochondrial 3 oxoacyl CoA thiolase|Mitochondrial 3 oxoacyl Coenzyme A thiolase|Mitochondrial 3-oxoacyl-CoA thiolase|T1|THIM_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42765
- P42765
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together