Order Entry
United States
ContactUsLinkComponent
Anti-MAP2K5 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MAP2K5 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MAP2K5 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77822-849
  • 77822-850
  • Antigen Symbol:
  • MAP2K5
  • MAP2K5
  • Antigen Name:
  • Dual specificity mitogen-activated protein kinase kinase 5 protein (1-190AA)
  • Dual specificity mitogen-activated protein kinase kinase 5 protein (1-190AA)
  • Antigen Synonyms:
  • Dual specificity mitogen activated protein kinase kinase 5|Dual specificity mitogen-activated protein kinase kinase 5|EC 2.7.12.2|HsT17454|MAP kinase kinase 5|MAP kinase kinase MEK5b|MAP2K5|MAPK/ERK kinase 5|MAPKK 5|MAPKK5|MEK 5|mitogen-activated protein kinase kinase 5|MKK5|MP2K5_HUMAN|PRKMK5|Protein kinase, mitogen-activated, kinase 5|SAPKK5|SKK5
  • Dual specificity mitogen activated protein kinase kinase 5|Dual specificity mitogen-activated protein kinase kinase 5|EC 2.7.12.2|HsT17454|MAP kinase kinase 5|MAP kinase kinase MEK5b|MAP2K5|MAPK/ERK kinase 5|MAPKK 5|MAPKK5|MEK 5|mitogen-activated protein kinase kinase 5|MKK5|MP2K5_HUMAN|PRKMK5|Protein kinase, mitogen-activated, kinase 5|SAPKK5|SKK5
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q13163
  • Q13163
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together