Anti-SLC38A2 Rabbit Polyclonal Antibody
77806-247
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of SNAT2.
- Antigen Symbol:SLC38A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96QD8
- Antigen Synonyms:Amino acid transporter 2|Amino acid transporter A2|ATA2|KIAA1382|PRO1068|Protein 40-9-1|S38A2_HUMAN|SAT2|Slc38a2|SNAT2|Sodium-coupled neutral amino acid transporter 2|Solute carrier family 38 member 2|System A amino acid transporter|System A amino acid transporter 2|System A transporter 1|System N amino acid transporter 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: SLC38A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human