Anti-UBASH3A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBASH3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-820
- 77811-820
- Antigen Symbol:
- UBASH3A
- UBASH3A
- Antigen Name:
- Ubiquitin-associated and SH3 domain-containing protein A protein (287-455AA)
- Ubiquitin-associated and SH3 domain-containing protein A protein (287-455AA)
- Antigen Synonyms:
- Cbl interacting protein 4|Cbl-interacting protein 4|CLIP4|STS 2|STS-2|Suppressor of T cell receptor signaling 2|Suppressor of T-cell receptor signaling 2|T cell ubiquitin ligand|T cell ubiquitin ligand protein|T-cell ubiquitin ligand 1|TULA|TULA-1|UBASH3A|ubiquitin associated and SH3 domain containing, A|Ubiquitin-associated and SH3 domain-containing protein A|UBS3A_HUMAN
- Cbl interacting protein 4|Cbl-interacting protein 4|CLIP4|STS 2|STS-2|Suppressor of T cell receptor signaling 2|Suppressor of T-cell receptor signaling 2|T cell ubiquitin ligand|T cell ubiquitin ligand protein|T-cell ubiquitin ligand 1|TULA|TULA-1|UBASH3A|ubiquitin associated and SH3 domain containing, A|Ubiquitin-associated and SH3 domain-containing protein A|UBS3A_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57075
- P57075
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together