Anti-HLA-DOA Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HLA-DOA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77817-004
- 77817-005
- Antigen Symbol:
- HLA-DOA
- HLA-DOA
- Antigen Name:
- HLA class II histocompatibility antigen, DO alpha chain protein (26-135AA)
- HLA class II histocompatibility antigen, DO alpha chain protein (26-135AA)
- Antigen Synonyms:
- HLA-DOA|HLA-DNA|HLA-DZAHLA class II histocompatibility antigen|DO alpha chain|MHC DN-alpha|MHC DZ alpha|MHC class II antigen DOA
- HLA-DOA|HLA-DNA|HLA-DZAHLA class II histocompatibility antigen|DO alpha chain|MHC DN-alpha|MHC DZ alpha|MHC class II antigen DOA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06340
- P06340
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together