Anti-PTTG1 Rabbit Polyclonal Antibody
Catalog # 77796-192
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TUTR1
- Antigen Symbol:PTTG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:O95997
- Antigen Synonyms:AW555095|C87862|Cut2|EAP 1|EAP1|ESP1 associated protein 1|Esp1-associated protein|hPTTG|MGC126883|MGC138276|Pds1|Pituitary tumor transforming 1|Pituitary tumor transforming protein 1|Pituitary tumor-transforming 1|isoform CRA_a|isoform CRA_b|Pituitary tumor-transforming gene 1|Pituitary tumor-transforming gene 1 protein|PTTG 1|PTTG|PTTG1|PTTG1 protein|PTTG1_HUMAN|Pttg3|Securin|Tumor transforming 1|Tumor transforming protein 1|Tumor-transforming protein 1|TUTR 1|TUTR1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PTTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat