Anti-Botulinum neurotoxin type C1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Botulinum neurotoxin type C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Clostridium botulinum
Specifications
- Catalog No:
- 77815-599
- 77815-598
- Antigen Symbol:
- Botulinum neurotoxin type C1
- Botulinum neurotoxin type C1
- Antigen Name:
- Recombinant Clostridium botulinum Botulinum neurotoxin type C1 protein (2-428AA)
- Recombinant Clostridium botulinum Botulinum neurotoxin type C1 protein (2-428AA)
- Antigen Synonyms:
- Botulinum neurotoxin type C|BoNT/C|Bontoxilysin-C1|BoNT/C1|Botulinum neurotoxin type C1) [Cleaved into: Botulinum neurotoxin C light chain|LC|EC 3.4.24.69)|Botulinum neurotoxin C heavy chain|HC)]
- Botulinum neurotoxin type C|BoNT/C|Bontoxilysin-C1|BoNT/C1|Botulinum neurotoxin type C1) [Cleaved into: Botulinum neurotoxin C light chain|LC|EC 3.4.24.69)|Botulinum neurotoxin C heavy chain|HC)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Clostridium botulinum
- Clostridium botulinum
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P18640
- P18640
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together